Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YM 750, Tocris Bioscience™

Catalog No. 303910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-391-0 10 mg
30-395-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-391-0 Supplier R&D Systems Supplier No. 3039/10
This item is not returnable. View return policy

ACAT inhibitor

Acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor (IC50 = 0.18 μM). Exhibits hypocholesterolaemic and antiatherosclerotic activity in vivo.

Chemical Identifiers

CAS 138046-43-2
Molecular Formula C31H36N2O
Molecular Weight (g/mol) 452.642
InChI Key FMLJREWZCZHGGW-UHFFFAOYSA-N
Synonym d0d9ad, acat inhibitors, yamanouchi, 1-cycloheptyl-1-9h-fluoren-2-ylmethyl-3-2,4,6-trimethylphenyl urea, n-cycloheptyl-n-9h-fluoren-2-ylmethyl-n'-2,4,6-trimethylphenyl urea
PubChem CID 9911716
IUPAC Name 1-cycloheptyl-1-(9H-fluoren-2-ylmethyl)-3-(2,4,6-trimethylphenyl)urea
SMILES CC1=CC(=C(C(=C1)C)NC(=O)N(CC2=CC3=C(C=C2)C4=CC=CC=C4C3)C5CCCCCC5)C

Specifications

Quantity 10 mg
Formula Weight 452.63
Percent Purity >98%
Chemical Name or Material YM 750
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.