Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ymer Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA573001
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA573001 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA573001 Supplier Invitrogen™ Supplier No. PA573001
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ymer Polyclonal Antibody for Western Blot

This protein is a bifunctional mRNA-capping enzyme exhibiting RNA 5'-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5'-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5'-diphosphate terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ymer
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene Ccdc50
Gene Accession No. Q8IVM0
Gene Alias C3orf6; C3orf6h; Ccdc50; coiled-coil domain containing 50; coiled-coil domain-containing protein 50; DFNA44; Protein Ymer; putative C3orf6 protein homolog; YMER; Ymer protein
Gene Symbols Ccdc50
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal portion of human Chromosome 3 open reading frame 6 (YMER).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152137
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.