Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YNB w/ Ammonium Sulfate w/o Folic Acid w/o PABA

Catalog No. MP114030112 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
227 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114030112 227 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114030112 Supplier MP Biomedicals™ Supplier No. 114030112
Add to Cart
Add to Cart

For use with Yeast Cell Culture and Gene Expression applications

Components Per liter mediumTrace ElementsBoric Acid 0.5 mgCopper Sulfate 0.04 mgPotassium Iodide 0.1 mgFerric Chloride 0.2 mgManganese Sulfate 0.4 mgSodium Molybdate 0.2 mgZinc Sulfate 0.4 mgSaltsMagnesium Sulfate 500 mgSodium Chloride 100 mgPotassium Phosphate, Monobasic 1000 mgCalcium Chloride 132 mgAmmonium SulfateVitamins5000 mgRiboflavin 0.2 mgBiotin 0.002 mgInositol 2 mgNicotinic Acid 0.4 mgPanthotenic Acid HemiCalcium Salt 0.4 mgPyridoxine Hydrochloride 0.4 mgThiamine Hydrochloride 0.4 mg
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Form Powder
Media Type Yeast
Product Type Microbiology Media
Quantity 227 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.