Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YOD1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734877 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4877 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4877 Supplier Proteintech Group Inc Supplier No. 253701AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Deubiquitinating enzymes (DUBs; see MIM 603478) areproteases that specifically cleave ubiquitin (MIM 191339) linkages,negating the action of ubiquitin ligases. DUBA8 belongs to a DUBsubfamily characterized by an ovarian tumor (OTU) domain.

Specifications

Antigen YOD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene YOD1
Gene Accession No. Q5VVQ6
Gene Alias DUBA 8, DUBA8, HIN 7, HIN7, HIV 1 induced protease 7, HsHIN7, OTUD2, PRO0907, Ubiquitin thioesterase OTU1, YOD1
Gene Symbols YOD1
Host Species Rabbit
Immunogen YOD1 Fusion Protein Ag21881
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55432
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.