Learn More
Medchemexpress LLC HY-18723 10mg , Yoda 1 CAS:448947-81-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1872310MG
Encompass_Preferred
HY-18723 10mg Yoda 1 CAS: 448947-81-7 Yoda 1 is a potent and selective Piezo1 agonist. Yoda 1 activates purified Piezo1 channels. Yoda1 induces activation of both Akt and ERK1/2 in endothelial cells (ECs), which is not dependent on Piezo1. Yoda 1 potently inhibits macropinocytosis induced by epidermal growth factor (EGF). Yoda 1 enhances Ca2+ influx followed by activation of the calcium-activated potassium channel KCa3.1 and inhibition of Rac1 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1025