Learn More
Selleck Chemical LLC Trimetazidine dihydrochloride 100mg 13171-25-0 Yoshimilon, Kyurinett, Vastarel F
SDP
Supplier:  Selleck Chemical LLC S4543100MG
Encompass_Preferred
Trimetazidine dihydrochloride (Yoshimilon, Kyurinett, Vastarel F) is a kind of drug for treatment of chronic ischemic disorders. It improves myocardial glucose utilization through inhibition of fatty acid metabolism, known as fatty acid oxidation inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6280