Learn More
Youngstown Barrel & Drum Ultra-Pipe Drip Diverter™

Description
Includes
Bungee cords and hooks for quick installation
- Keep fluid leaks off the floor
- Eliminate slip hazards and chemical exposure
- Large-diameter opening captures overhead leaks, big and small
- Bottom fitting connects to standard garden hose, allowing fluids to be collected in drum or tank
- Rugged, coated vinyl construction
- Dimensions: 18 in.D × 18 in. H (45.7 × 45.7cm)
- Weight: 2 lb. (0.91kg)
- Material: 18 oz. coated vinyl
- Capacity: 7 gal. (26.5L)
Specifications
Specifications
| Capacity (English) | 7 gal. |
| Capacity (Metric) | 26.5 L |
| Material | Vinyl |
| Type | Drip Diverter |
| For Use With (Application) | Keep fluid leaks off the floor to eliminate slip hazards and chemical exposure, captures overhead leaks, bottom fitting connects to a standard garden hose, allows fluids to be collected in a drum or tank |
| For Use With (Equipment) | Channel pipes |
| Includes | Bungee cords and hooks for quick installation |
| Packaging Quantity | 1 Ea. |
| Length (English) | 18 in. |
| Length (Metric) | 45.72 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.