Learn More
Youngstown Barrel & Drum Accessory Parts for Open-Head Steel and Plastic Drums

Description
Accessory Parts for Open-Head Steel and Plastic Drums. New Steel Bolt Ring 55 Gal 12Ga Drop Forged Lug.
Specifications
Specifications
| Product Type | Drum Lid (Plain) |
| Capacity (Metric) | 208 L |
| Color | Black |
| For Use With (Equipment) | 55 gal. Open Head Steel and Plastic Drums |
| Material | Steel (18 ga) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.