Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Containment Berm Ground Tarps
SDP

Catalog No. 19120093
Change view
Click to view available options
Length (English):
54 ft.
70 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-120-093 54 ft.
19-120-091 70 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-120-093 Supplier Youngstown Barrel & Drum Supplier No. 8338 UT

Place under berm to protect it from hazards that may damage it.

Containment Berm Ground Tarp. 19' x 54' For all 15' x 50' Containment BermsThis item go underneath the Berm and are typically 1 to 4 feet larger than the Berm itself. It is made from a 18.5 oz/sq. yd. polyester blended PVC.

Specifications

Color Black
Material Polyester-blended PVC
Type Ground Tarp
For Use With (Application) Protect the berm materials from punctures, tears and cuts that might occur from stones, glass, and other debris
For Use With (Equipment) All 15 x 50 ft. Containment Berms
Model Ground Tarp
Packaging Quantity 1 Ea.
Shape Rectangular
Dimensions (L x W) 54 x 19 ft. (16.45 x 5.79 m)
Length (English) 54 ft.
Length (Metric) 16.45 m
Width (English) 19 ft.
Width (Metric) 5.8 m
Weight (English) 68 lb.
Weight (Metric) 30.8 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.