Learn More
Youngstown Barrel & Drum Ultra-1000 Containment Sumps™

Description
Extra-large sumps contains leaks, spills and overfills
- Help prevent groundwater contamination outside and slippery floors inside
- One-piece molded polyethylene design prevents rust and corrosion
- Nestable design saves freight and minimizes storage space
- Optional 1.9 or 5.08cm (0.75 or 2 in.) drain plug available for convenient emptying
- Great for indoor or outdoor oil tanks
Compliance
Meets stormwater, SPCC and EPA spill containment regulations
Specifications
Specifications
| Certifications/Compliance | 40 CFR 264.175, SPCC, EPA |
| Capacity (English) | 360 gal. |
| Capacity (Metric) | 1362.75 L |
| Material | Polyethylene |
| Type | Ultra-Containment Sump |
| For Use With (Application) | Eliminates costly spills while storing fuels, oil and other hazardous liquids by completely containing tanks and also for outdoor applications |
| For Use With (Equipment) | 275 gal. oval tank |
| Includes | Drain |
| Packaging Quantity | 1 Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.