Learn More
Youngstown Barrel & Drum Ultra-Containment Berms™ Stake Wall Models: Replacement Parts

Description
For Ultra-Containment Berms, see cat. no. 19339015 - 9
Specifications
Specifications
| Color | Yellow |
| Type | Replacement Stake |
| For Use With (Application) | Designed with a base hinge that allows the stake to move back and forth as items enter or exit the containment area |
| For Use With (Equipment) | Ultra-Containment berms |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.