Learn More
Youngstown Barrel & Drum Ultra-Containment Trays

Description
Compliance
Meets EPA Container Storage Regulations for small packages with grating in place
Specifications
Specifications
| Certifications/Compliance | Meets EPA Container Storage Regulations for small packages with grating in place. |
| Color | Yellow |
| Capacity (English) | 14 gal. |
| Capacity (Metric) | 53 L |
| Material | Polyethylene |
| Type | Ultra-Containment Tray |
| For Use With (Application) | Used as a storage pad for batteries and other hazardous packages, for small containers and packages |
| Includes | 2 x 4 ft. grating elevates containers |
| Load Bearing Capacity (English) | 3,000 lb. |
| Load Bearing Capacity (Metric) | 1360.8 kg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.