Learn More
Youngstown Barrel & Drum Ultra-Funnels™
SDP
Catalog No. 19820909
Change view
Click to view available options
Design:
Closed or open head
Horseshoe
Open head
Type:
Global Funnel
Snap-on Funnel Cover
Ultra-Open Head Funnel
Universal Funnel
For Use With (Equipment):
22.25 to 23.5 in. 55 gal. drum
55 and 30 gal. drums
55 gal. tight head steel or polyethylene drum
Includes:
Cover only
Funnel only
Hinged cover, spout
Snap-on cover
Spout
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Design Type For Use With (Equipment) Includes
19-820-909 Horseshoe Global Funnel 55 gal. tight head steel or polyethylene drum Hinged cover, spout
19-820-907 Closed or open head Universal Funnel 55 and 30 gal. drums Spout
19-130-4817 Horseshoe Global Funnel 55 gal. tight head steel or polyethylene drum Spout
19-130-4818 Horseshoe Global Funnel 55 gal. tight head steel or polyethylene drum Snap-on cover
19-130-4819 Open head Ultra-Open Head Funnel 22.25 to 23.5 in. 55 gal. drum Funnel only
19-130-4821 Closed or open head Snap-on Funnel Cover 55 and 30 gal. drums Cover only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-820-909 Supplier Youngstown Barrel & Drum Supplier No. 0499 UT
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Prevent spills and messes of all kinds with these high-quality funnels
Allows accurate monitoring of vessel filling
Global Funnels
  • Securely fits nearly every 55-gallon tighthead steel or polyethylene drum
  • Unique bottom design fits tightly on drums with varying diameters
  • No more wobbles or drips
Open Head Funnel
  • All-polyethylene funnel can be used on 55-gallon open-head steel or polyethylene drums
  • Replaces drum lid for convenient access for pouring, collecting rags, soiled absorbents, etc.
  • Molded base fits securely onto drum; thumb screws quickly tighten for added security
  • Hinged cover is lockable
Specifications
Design Horseshoe
Capacity (English) 5 gal.
Capacity (Metric) 18.92 L
Type Global Funnel
For Use With (Application) Allows installation of a PopUp fill gauge which allows accurate monitoring of liquid level inside the drum to prevent messy overfilling
For Use With (Equipment) 55 gal. tight head steel or polyethylene drum
Includes Hinged cover, spout
Packaging Quantity 1 Ea.
Weight (English) 18 lb.
Weight (Metric) 8.16 kg