Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Hard Top Plus™ Models
SDP

Catalog No. 19339037
Change view
Click to view available options
Weight (English):
815 lb.
1190 lb.
1565 lb.
1940 lb.
Dimensions (L x W x H):
126 x 62 x 79 in.
188 x 62 x 79 in.
250 x 62 x 79 in.
312 x 62 x 79 in.
Holds:
12 Drums
16 Drums
20 Drums
8 Drums
Includes:
Drain
No Drain
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English) Dimensions (L x W x H) Holds Includes
19-339-037 1940 lb. 312 x 62 x 79 in. 20 Drums No Drain
19-339-033 1190 lb. 188 x 62 x 79 in. 12 Drums No Drain
19-339-032 815 lb. 126 x 62 x 79 in. 8 Drums Drain
19-339-031 815 lb. 126 x 62 x 79 in. 8 Drums No Drain
19-339-034 1190 lb. 188 x 62 x 79 in. 12 Drums Drain
19-339-035 1565 lb. 250 x 62 x 79 in. 16 Drums No Drain
19-339-036 1565 lb. 250 x 62 x 79 in. 16 Drums Drain
19-339-038 1940 lb. 312 x 62 x 79 in. 20 Drums Drain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-339-037 Supplier Youngstown Barrel & Drum Supplier No. 9656 UT

Provide a safe, convenient and effective place to store drums

All-polyethylene construction offers excellent chemical resistance and will not rust or corrode.

  • Low-profile containment pallet positions drum-tops at a safe, convenient level to pour wastes into funnels or to use drum pumps
  • Multiple 4-drum compartments offer flexible usage for a broad range of operations including: storage of virgin chemicals, active hazardous waste collection and storage of spill response supplies
  • Internal walls and separate sumps allow materials to be segregated
  • Swing-out doors provide safety and convenience for loading / unloading drums
  • Roll-top covers can be easily lifted from waist height to access drum tops

Compliance

Meets EPA Container Storage Regulation 40 CFR 264.175 and helps comply with NPDES, 40 CFR 122.26 (1999). Also meets SPCC Regulations for Secondary Containment of hydrocarbons.

Specifications

Certifications/Compliance SPCC, EPA, 40 CFR 264.175, NPDES, 40 CFR 122.26
Color Yellow
Capacity (English) 55 gal.
Capacity (Metric) 208 L
Material Polyethylene
Holds 20 Drums
Includes No Drain
Model P20
Resistant Chemical
Dimensions (L x W x H) 312 x 62 x 79 in.
Weight (English) 1940 lb.
Weight (Metric) 880 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.