Learn More
Youngstown Barrel & Drum Ultra-Hard Top Plus™ Models

Description
All-polyethylene construction offers excellent chemical resistance and will not rust or corrode.
- Low-profile containment pallet positions drum-tops at a safe, convenient level to pour wastes into funnels or to use drum pumps
- Multiple 4-drum compartments offer flexible usage for a broad range of operations including: storage of virgin chemicals, active hazardous waste collection and storage of spill response supplies
- Internal walls and separate sumps allow materials to be segregated
- Swing-out doors provide safety and convenience for loading / unloading drums
- Roll-top covers can be easily lifted from waist height to access drum tops
Compliance
Meets EPA Container Storage Regulation 40 CFR 264.175 and helps comply with NPDES, 40 CFR 122.26 (1999). Also meets SPCC Regulations for Secondary Containment of hydrocarbons.
Specifications
Specifications
| Certifications/Compliance | SPCC, EPA, 40 CFR 264.175, NPDES, 40 CFR 122.26 |
| Color | Yellow |
| Capacity (English) | 55 gal. |
| Capacity (Metric) | 208 L |
| Material | Polyethylene |
| Holds | 20 Drums |
| Includes | Drain |
| Model | P20 |
| Resistant | Chemical |
| Dimensions (L x W x H) | 312 x 62 x 79 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.