Learn More
Youngstown Barrel & Drum Ultra-Hose Stand™
SDP
Catalog No. 19339056
Change view
Click to view available options
Material:
Polyethylene
Polyurea
Model:
Collapsible
Heavy-Duty
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Model
19-339-056 Polyurea Heavy-Duty
18-938-438 Polyethylene Collapsible
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-339-056 Supplier Youngstown Barrel & Drum Supplier No. 8369 UT
Keeps hoses off of berm sidewalls - helps maintain structural integrity and capacity of Containment Berm
  • Versatile unit that can be used completely inside or outside of Berm or straddling Berm with sidewall running through 8 in. wide opening in support sections
  • Heavy-duty, ribbed polyethylene support sections are compatible with fuel and many other chemicals
  • Unit quickly and easily disassembles into four flat pieces for easy transport and storage
  • Cutout in top of unit can handle up to 6.5 in. hoses and keeps them secure
  • Wide 27 in. dia. base keeps unit in place and upright
  • Dimensions: 27 × 27 × 23.875 in. (68.6 × 65.6 × 60.64cm)
Specifications
Material Polyurea
Type Ultra Hose Stand
Model Heavy-Duty