Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Pop Up Pools Plus™
SDP

Catalog No. 19171136
Change view
Click to view available options
Capacity (Metric):
75.7 L
249.8 L
567.8 L
Length (Metric):
45.72 cm
152.4 cm
365.7 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Length (Metric)
19-171-136 75.7 L 45.72 cm
19-171-138 567.8 L 365.7 cm
19-171-137 249.8 L 152.4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-171-136 Supplier Youngstown Barrel & Drum Supplier No. 8010 UT

Opens and is ready to respond in seconds - minimizes spillage and exposure of hazardous material to the environment

  • Spill Response Pool folds into compact package until needed - Quickly opens and bladder unfurls to capture large spills
  • Ultra-Pop Up Pool Plus is available in 150, 66 and 20 gal. sizes
  • Unfold pool and open Bladder Compartment - bladder will automatically expand/unfurl as liquid level in pool rises
  • Compact folded size - stores efficiently inside spill kits, under/behind seats or in tool boxes
  • Excellent chemical resistance, including diesel fuel, antifreeze, acids, caustics and corrosives
  • Helps minimize environmental damage and related clean-up costs
  • Excellent for response to damaged saddle tanks and cross-over lines, hazmat spills, leaking containers, machinery and piping

Specifications

Capacity (English) 20 gal.
Capacity (Metric) 75.7 L
Material Polyethylene
Type Spill Control Pool
For Use With (Application) Minimizes spillage and exposure of hazardous material to the environment
For Use With (Equipment) Saddle tanks and cross-over lines, hazmat spills, leaking containers, machinery and piping
Includes Unfold pool and open bladder compartment with bladder attachment
Dimensions (L x W x H) 18 x 58 x 5 in. (45.72 x 147.32 x 12.7 cm)
Length (English) 18 in.
Length (Metric) 45.72 cm
Width (English) 58 in.
Width (Metric) 147.32 cm
Height (English) 5 in.
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.