Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Rack Containment Tray™
SDP

Catalog No. 19171076
Change view
Click to view available options
Capacity (English):
8 gal.
16 gal.
24 gal.
Weight (English):
8 lb.
17 lb.
27 lb.
Width (English):
23.5 in.
48 in.
72 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Weight (English) Width (English)
19-171-076 8 gal. 8 lb. 23.5 in.
19-171-078 24 gal. 27 lb. 72 in.
19-171-077 16 gal. 17 lb. 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-171-076 Supplier Youngstown Barrel & Drum Supplier No. 2370 UT
This item is not returnable. View return policy

Capture leaks and spills underneath pallet racking

  • Low-profile trays slide easily under pallet racking and shelving
  • Capture small leaks and spills from palletized contents above
  • Keep floors clean and free from slip hazards
  • Provide excellent chemical compatibility and corrosion resistance
  • Multiple-tray systems available to fit any size rack or shelf
  • Additional trays and/or connectors may be added for larger applications or revised configurations
  • Black, all-polyethylene construction

Specifications

Color Black
Design Low profile
Capacity (English) 8 gal.
Capacity (Metric) 30.28 L
Material Polyethylene
Type Spill Containment Tray
For Use With (Application) Capture small leaks and spills from palletized contents, keeping floors clean and free from slip hazard, provides excellent chemical compatibility and corrosion resistance
For Use With (Equipment) Any size rack or shelf
Includes Single tray
Packaging Quantity 1 Ea.
Dimensions (L x W x H) 44 x 23.5 x 2.75 in. (111.76 x 59.69 x 6.98 cm)
Length (English) 44 in.
Length (Metric) 111.76 cm
Width (English) 23.5 in.
Width (Metric) 59.69 cm
Height (English) 2.75 in.
Height (Metric) 6.98 cm
Weight (English) 8 lb.
Weight (Metric) 3.5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.