Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Recycled Overpack 95
SDP

Catalog No. 19171355
Change view
Click to view available options
Capacity (English):
95 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-171-355 95 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-171-355 Supplier Youngstown Barrel & Drum Supplier No. 0570 UT

Made from a low density polyethylene. It offers excellent chemical resistance against acids, caustics and corrosives. Applications: Overpacking of damaged or leaking packages. Collection and transport of soiled sorbents.

Ultra-Recycled Overpack 95 is the highest performing overpack poly drum on the market. Constructed from a low density polyethylene, it offers excellent chemical resistance against acids, caustics and corrosives. Applications: Overpacking of damaged or leaking packages. Collection and transport of soiled sorbents. Contaminated sites clean up. Emergency response. Direct containment and transport of hazardous solids. Spill kits. This Overpack is made with 100% recycled materials. Certified to be used as a Salvage Drum by the DOT under 49 CFR 173.3 (c). It also complies with UN Packaging Group 2 (Y-Rating).

Specifications

Certifications/Compliance UN, 1H2/Y345/S/11/USA/M5904, 49 CFR 173.3
Color Black
Capacity (English) 95 gal.
Capacity (Metric) 360 L
Material Recycled Materials
Type Salvage/Overpack drum
For Use With (Application) Overpacking of damaged or leaking packages
Includes Screw top lid
Model Overpack
Packaging Quantity 1 Ea.
Thickness 0.145 in.
Dimensions (L x W x H) 32.25 x 32.25 x 41.25 in.
Length (English) 32.25 in.
Length (Metric) 820 cm
Width (English) 32.25 in.
Width (Metric) 820 cm
Height (English) 41.25 in.
Height (Metric) 1049 cm
Depth (English) 37 in.
Depth (Metric) 93.98 cm
Diameter (English) 32.25 in.
Diameter (Metric) 82.5 cm
Weight (English) 48 lb.
Weight (Metric) 21.7 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.