Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Sidewinder™ Cable Protection System: 1 Ft. Extension System
SDP

Catalog No. 18938525
Change view
Click to view available options
Color:
Black
Black/Yellow
Size:
Medium
Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
18-938-525 Black/Yellow Large
18-938-521 Black/Yellow Medium
18-938-523 Black Medium
18-938-527 Black Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-938-525 Supplier Youngstown Barrel & Drum Supplier No. 1841 UT

For use with Ultra-Sidewinder Cable Protection System

One-foot extension. An innovative new solution for protecting cables run across a floor. The new design articulates left and right, allowing the system to turn corners and adapt to the layout of the room, while maintaining full protection for the cables and trip protection for pedestrians. Cables can be left in the system for the ultimate in portability, or left in place indefinitely.

Available in medium and large sizes in black or black-and-yellow

Specifications

Product Type Ultra-Sidewinder Cable Protection System
Size Large
Color Black/Yellow
Dimensions (L x W x H) 34 x 13-5/8 x 2-1/8 in.
Height (English) 1.5 in.
Height (Metric) 38 mm
Style 1-Foot Extension System
Weight (English) 13.5 lb.
Weight (Metric) 6 kg
Width (English) 13-5/8 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.