Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Sidewinder™ Cable Protection System: 3 Ft.
SDP

Catalog No. 18938526
Change view
Click to view available options
Color:
Black
Black/Yellow
Size:
Medium
Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
18-938-526 Black Large
18-938-520 Black/Yellow Medium
18-938-522 Black Medium
18-938-524 Black/Yellow Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-938-526 Supplier Youngstown Barrel & Drum Supplier No. 1842 UT

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Protects cords and cables and avoid dangerous tripping hazards

An innovative new solution for protecting cables run across a floor. The new design articulates left and right, allowing the system to turn corners and adapt to the layout of the room, while maintaining full protection for the cables and trip protection for pedestrians. Cables can be left in the system for the ultimate in portability, or left in place indefinitely.

  • Articulating pieces offer flexibility to design curves, corners and s-curves while maintaining completely flat profile
  • Modular design enables reconfigure, repair, and replace targeted segments, even while the cable remains connected.
  • Simple snap in assembly without cutting or using tools
  • Fully customizable length
  • Black and yellow color provides additional safety tape type visual warning
  • Also available in all black
  • Load Capacity: 260 lbs./ in.2 (18.3 kg./cm2)
  • Load Capacity per axle: 13,500 lbs. (6,123 kg)
  • Available in medium and large sizes
  • One-foot extensions available separately

Specifications

Product Type Ultra-Sidewinder Cable Protection System
Size Large
Color Black
Height (English) 1.5 in.
Height (Metric) 38 mm
Style 3-Foot System
Weight (English) 13.5 lb.
Weight (Metric) 6 kg
Width (English) 3 in.
Width (Metric) 76 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.