Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Spill Decks™
SDP

Catalog No. 19058033
Change view
Click to view available options
Length (English):
24 in.
25.9 in.
52 in.
Load Bearing Capacity (English):
600 lb.
1500 lb.
3000 lb.
6000 lb.
Weight (English):
17 lb.
23 lb.
40 lb.
70 lb.
Width (English):
25.9 in.
32 in.
52 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Load Bearing Capacity (English) Weight (English) Width (English)
19-058-033 24 in. 600 lb. 17 lb. 32 in.
19-058-075 52 in. 3000 lb. 40 lb. 25.9 in.
19-058-043 25.9 in. 1500 lb. 23 lb. 25.9 in.
19-058-074 52 in. 6000 lb. 70 lb. 52 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-058-033 Supplier Youngstown Barrel & Drum Supplier No. 1089 UT

Allow you to design and create a containment system that meets your specific needs

Applications include hazardous waste collection, storage of virgin chemicals, drum dispensing, and battery storage. Perfect for satellite collection of hazardous wastes.

  • Polyethylene construction meets a wide range of chemical compatibility needs
  • Bulkhead fittings lock connected modules together and allow spilled liquid to flow from one module to the next
  • Low profile (5.75 in.) height offers safe and convenient loading/unloading of drums
  • Easy to assemble, no tools required

Order Info

Optional loading ramp provides added drum handling safety.

Compliance

Meet EPA and Uniform Fire Code Spill Containment Regulations when assembled in configurations of six or more. Meet EPA Container Storage Regulation 40CFR264.175.

Specifications

Certifications/Compliance EPA, 40 CFR 264.175
Color Yellow
Material Polyethylene
Type Loading Ramp
For Use With (Application) Hazardous waste collection, storage of virgin chemicals, drum dispensing, and battery storage, perfect for satellite collection of hazardous wastes
For Use With (Equipment) Spill decks
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 272 kg
Dimensions (L x W x H) 24 x 32 x 5.75 in. (60.96 x 81.3 x 14.6 cm)
Length (English) 24 in.
Length (Metric) 60.96 cm
Width (English) 32 in.
Width (Metric) 81.3 cm
Height (English) 5.75 in.
Height (Metric) 14.6 cm
Weight (English) 17 lb.
Weight (Metric) 7.7 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.