Learn More
Youngstown Barrel & Drum Ultra-Spill Decks™

Description
Applications include hazardous waste collection, storage of virgin chemicals, drum dispensing, and battery storage. Perfect for satellite collection of hazardous wastes.
- Polyethylene construction meets a wide range of chemical compatibility needs
- Bulkhead fittings lock connected modules together and allow spilled liquid to flow from one module to the next
- Low profile (5.75 in.) height offers safe and convenient loading/unloading of drums
- Easy to assemble, no tools required
Order Info
Optional loading ramp provides added drum handling safety.
Compliance
Meet EPA and Uniform Fire Code Spill Containment Regulations when assembled in configurations of six or more. Meet EPA Container Storage Regulation 40CFR264.175.
Specifications
Specifications
| Certifications/Compliance | EPA, 40 CFR 264.175 |
| Color | Yellow |
| Capacity (English) | 11 gal. |
| Capacity (Metric) | 41.6 kg |
| Material | Polyethylene |
| Type | P1 Module Ultra-Spill Deck |
| For Use With (Application) | Hazardous waste collection, storage of virgin chemicals, drum dispensing, and battery storage, perfect for satellite collection of hazardous wastes |
| Holds | 1 Drum |
| Load Bearing Capacity (English) | 1500 lb. |
| Load Bearing Capacity (Metric) | 680 kg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.