Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Spill Pallet; Accessories: Pullover Cover
SDP

Catalog No. 191304988
Change view
Click to view available options
For Use With (Application):
Keeps rainwater out of sump and helps comply with stormwater management regulations
Protects drums and pallets from weather elements
For Use With (Equipment):
4-drum spill containment pallet with drums
IBC spill pallet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) For Use With (Equipment)
19-130-4988 Keeps rainwater out of sump and helps comply with stormwater management regulations IBC spill pallet
19-820-913 Protects drums and pallets from weather elements 4-drum spill containment pallet with drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-130-4988 Supplier Youngstown Barrel & Drum Supplier No. 1055 UT

Cover items placed on the P4 Spill Pallet.

Ultra-Spill Pallet Pullover Cover: Accessories. Keeps rainwater out of sump and helps comply with stromawater management regulations.

Specifications

Certifications/Compliance SPCC, EPA
Color Yellow
Material Polyester
Type Pull Over Cover
For Use With (Application) Keeps rainwater out of sump and helps comply with stormwater management regulations
For Use With (Equipment) IBC spill pallet
Packaging Quantity 1 Ea.
Resistant Mildew, Flame (Ul-94, V-0, NFPA-701 TM 1)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.