Learn More
Youngstown Barrel & Drum Ultra-Spill Pallets Plus™

Description
- Nearly 50% lower than standard spill pallets for safer and less strenuous handling
- Removable grates feature non-slip surface
- All-polyethylene construction
- Forkliftable
Order Info
Available in one-, two-, three- and four-drum configurations, with or without a drain
Compliance
Meet U.S. EPA container storage regulation 40 CFR 264.175, Uniform Fire Code spill containment regulations for use with drums and is SPCC compliant.
Specifications
Specifications
| Certifications/Compliance | SPCC, EPA, 40 CFR 264.175, UFC |
| Color | Yellow |
| Material | Vinyl |
| Type | Pull Over Cover |
| For Use With (Application) | Reduces the potential for hazardous fluids to splash into the eyes or face |
| For Use With (Equipment) | P2 Ultra-spill Pallet™ plus |
| Includes | Cover |
| Model | Cover |
| Weight (English) | 4 lb. |
| Weight (Metric) | 1.81 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.