Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Spill Pallets Plus™
SDP

Catalog No. 19171079
Change view
Click to view available options
Capacity (English):
62 gal.
66 gal.
75 gal.
Type:
Pull Over Cover
Ultra-Spill Pallet P1 Plus
Ultra-Spill Pallet P2 Plus
Ultra-Spill Pallet P4 Plus
Includes:
Cover
Drain
No drain
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Type Includes
19-171-079 62 gal. Ultra-Spill Pallet P1 Plus No drain
19-130-4780 75 gal. Ultra-Spill Pallet P4 Plus Drain
19-130-5027 66 gal. Ultra-Spill Pallet P2 Plus No drain
18-938-502 - Pull Over Cover Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-171-079 Supplier Youngstown Barrel & Drum Supplier No. 9606 UT

Low-profile design—only 8.75 in. high (22.2cm)

  • Nearly 50% lower than standard spill pallets for safer and less strenuous handling
  • Removable grates feature non-slip surface
  • All-polyethylene construction
  • Forkliftable

Order Info

Available in one-, two-, three- and four-drum configurations, with or without a drain

Compliance

Meet U.S. EPA container storage regulation 40 CFR 264.175, Uniform Fire Code spill containment regulations for use with drums and is SPCC compliant.

Specifications

Certifications/Compliance SPCC, EPA, 40 CFR 264.175, UFC
Color Yellow
Capacity (English) 62 gal.
Capacity (Metric) 234.6 L
Material Polyethylene
Type Ultra-Spill Pallet P1 Plus
For Use With (Application) Reduces the potential for hazardous fluids to splash into the eyes or face
Holds 1 Drum
Includes No drain
Load Bearing Capacity (English) 800 lb.
Load Bearing Capacity (Metric) 362.8 kg
Model P1
Dimensions (L x W x H) 40 x 40 x 12 in. (101.6 x 101.6 x 30.48 cm)
Length (English) 40 in.
Length (Metric) 101.6 cm
Width (English) 40 in.
Width (Metric) 101.6 cm
Height (English) 12 in.
Height (Metric) 30.48 cm
Weight (English) 51 lb.
Weight (Metric) 23 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.