Learn More
Youngstown Barrel & Drum Ultra Track Belts

Description
Track belts are designed to be used with ultra-containment berms.
- Material lies inside of containment berm
- Runs the length of the berm's usable space
- Acts as a buffer between vehicle tires and berm
- All-polyester, single-ply interwoven carcass
- Low stretch and good tear resistance
- Color: Yellow and black
Specifications
Specifications
| Color | Yellow/Black |
| Material | PVC |
| Type | Track Belt Set |
| For Use With (Application) | Provide additional support between vehicles tires and containment berm surface |
| For Use With (Equipment) | Economy containment berm |
| Dimensions (L x W x H) | 60 x 30 x 0.062 in. (1524 x 76.2 x 0.15 cm) |
| Length (English) | 60 ft. |
| Length (Metric) | 1524 cm |
| Width (English) | 30 in. |
| Width (Metric) | 76.2 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.