Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra Track Belts
SDP

Catalog No. 191305007
Change view
Click to view available options
Length (English):
50 ft.
60 ft.
66 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-130-5007 66 ft.
19-130-5005 50 ft.
19-130-5006 60 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-130-5007 Supplier Youngstown Barrel & Drum Supplier No. 8345 UT

Heavy-duty, reinforced PVC compound offers excellent oil/hydrocarbon resistance and long wear

Track belts are designed to be used with ultra-containment berms.

  • Material lies inside of containment berm
  • Runs the length of the berm's usable space
  • Acts as a buffer between vehicle tires and berm
  • All-polyester, single-ply interwoven carcass
  • Low stretch and good tear resistance
  • Color: Yellow and black

Specifications

Color Yellow/Black
Material PVC
Type Track Belt Set
For Use With (Application) Provide additional support between vehicles tires and containment berm surface
For Use With (Equipment) Economy containment berm
Dimensions (L x W x H) 66 x 30 x 0.062 in. (2011 x 76.2 x 0.15 cm)
Length (English) 66 ft.
Length (Metric) 2011 cm
Width (English) 30 in.
Width (Metric) 76.2 cm
Height (English) 0.062 in.
Height (Metric) 0.15 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.