Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Track Pans™
SDP

Catalog No. 19120139
Change view
Click to view available options
Type:
Bulkhead Fitting
Left Grate
Railroad Spill Containment System
Right Grate
Ultra-Track Center Pan
Ultra-Track Side Pan
For Use With (Application):
Below grade piping
Capture hazardous spills from railroad tanker cars, to provide spill containment at industrial rail sidings and locomotive maintenance facilities
Provide spill containment at industrial rail sidings and locomotive maintenance facilities
For Use With (Equipment):
Railroad tanker cars
Ultra-track center pan
Ultra-track pans with grates
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Application) For Use With (Equipment)
19-120-139 Ultra-Track Side Pan Provide spill containment at industrial rail sidings and locomotive maintenance facilities Railroad tanker cars
19-120-137 Ultra-Track Center Pan Provide spill containment at industrial rail sidings and locomotive maintenance facilities Railroad tanker cars
19-120-144 Railroad Spill Containment System Capture hazardous spills from railroad tanker cars, to provide spill containment at industrial rail sidings and locomotive maintenance facilities Railroad tanker cars
19-120-147 Railroad Spill Containment System Capture hazardous spills from railroad tanker cars, to provide spill containment at industrial rail sidings and locomotive maintenance facilities Railroad tanker cars
19-120-150 Railroad Spill Containment System Capture hazardous spills from railroad tanker cars, to provide spill containment at industrial rail sidings and locomotive maintenance facilities Railroad tanker cars
19-130-5016 Right Grate Provide spill containment at industrial rail sidings and locomotive maintenance facilities Ultra-track center pan
19-130-5015 Left Grate Provide spill containment at industrial rail sidings and locomotive maintenance facilities Ultra-track center pan
19-130-5013 Bulkhead Fitting Below grade piping Ultra-track pans with grates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-120-139 Supplier Youngstown Barrel & Drum Supplier No. 9576 UT

Includes

  • Center track pan: 2 bulkhead fittings, 4 gaskets and 2 grates
  • Side track pan: 1 bulkhead fittings, 2 gaskets and 1 grates

Designed to provide railroad spill containment at industrial rail sidings and locomotive maintenance facilities.

Available in a range of lengths, Ultra-Track Pans may be used to collect small spills and leaks or to capture and channel off significant spills caused by defective equipment or a major overflow.

  • Prevent costly cleanup and regulatory fines from railcar spills and chemical spills
  • Railroad spill containment areas can be easily assembled to any length desired
  • Rugged all-polyethylene construction withstands harsh chemicals and degrading effects of the sun
  • Polyethylene gaskets keep spills from falling between the rails and pans
  • Trucks and other vehicles can drive over track pans with grates in place
  • Designed to function in temperatures ranging from -40°F to +140°F

  • Optional track pan covers keep rainwater out of the pans, providing effective stormwater management
  • Lightweight track pans can be easily installed without heavy lifting equipment, e.g., forklifts, cranes, or backhoes, etc.
  • Pans, grates and covers are designed with slip resistant surfaces
  • Estimated to perform 20–30 years; longer than fiberglass pans

9–Foot Railroad Spill Containment Systems:

  • 2 center pans and 4 side pans, with or without covers
  • 40–Foot Railroad Spill Containment Systems:
    • 9 center pans and 18 side pans, with or without covers
  • 60–Foot Railroad Spill Containment Systems:
    • 14 center pans and 28 side pans, with or without covers

    Compliance

    Meets SPCC and EPA Stormwater Management Regulations; meets New York State Chemical Bulk Storage Regulation Section 599.17c – Secondary Containment for Transfer Stations

    Specifications

    Certifications/Compliance 40 CFR 264.175, SPCC, EPA
    Capacity (English) 19 gal.
    Capacity (Metric) 71.9 L
    Material Polyethylene
    Type Ultra-Track Side Pan
    For Use With (Application) Provide spill containment at industrial rail sidings and locomotive maintenance facilities
    For Use With (Equipment) Railroad tanker cars
    Includes 1 bulkhead fitting, 12 oz. can, urethane Foam, 1 gasket, 1 grate
    Packaging Quantity 1 Ea.
    Temperature Range (English) -40° to 140°F
    Temperature Range (Metric) -40° to 60°C
    Dimensions (L x W x H) 53.5 x 27.5 x 6 in. (135.89 x 69.85 x 15.24 cm)
    Length (English) 53.5 in.
    Length (Metric) 135.89 cm
    Width (English) 27.5 in.
    Width (Metric) 69.85 cm
    Height (English) 6 in.
    Height (Metric) 15.24 cm
    Weight (English) 132.27 lb.
    Weight (Metric) 60 kg
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.