Learn More
Youngstown Barrel & Drum UT Ultra-Curb Guard™, Insert-Style Replacement Filter
SDP
Catalog No. 191305002
Change view
Click to view available options
For Use With (Equipment):
9760 curb guard
9761 curb guard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-130-5002 9760 curb guard
19-130-5003 9761 curb guard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-130-5002 Supplier Youngstown Barrel & Drum Supplier No. 9762 UT
Keeps oil, sediment and floatables out of curb inlets
  • For use in drains with curb inlets only
  • Two heavy-duty tension rods
  • Features X-Tex filtering material
Specifications
Material X-Tex
Type Replacement Filter
For Use With (Application) Use in drains with curb inlets only, X-Tex filtering material
For Use With (Equipment) 9760 curb guard