Learn More
Youngstown Barrel & Drum UT Ultra-Curb Guard™, Insert-Style
Description
- For use in drains with curb inlets only
- Two heavy-duty tension rods
- Features X-Tex filtering material
Specifications
Specifications
| Certifications/Compliance | NPDES, 40 CFR 122.26, TMDL |
| Material | X-Tex |
| Type | Ultra Curb Guard Insert |
| For Use With (Application) | Keep oil, sediment, floatables and other contaminants out of curb inlets |
| For Use With (Equipment) | 22 to 35 in. inlets |