Learn More
Youngstown Barrel & Drum UT Ultra-Curb Guard™, Insert-Style
SDP
Catalog No. 191305001
Change view
Click to view available options
For Use With (Equipment):
22 to 35 in. inlets
36 to 60 in. inlets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-130-5001 36 to 60 in. inlets
19-130-5000 22 to 35 in. inlets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-130-5001 Supplier Youngstown Barrel & Drum Supplier No. 9761 UT
Keeps oil, sediment and floatables out of curb inlets
  • For use in drains with curb inlets only
  • Two heavy-duty tension rods
  • Features X-Tex filtering material
Specifications
Certifications/Compliance NPDES, 40 CFR 122.26, TMDL
Material X-Tex
Type Ultra Curb Guard Insert
For Use With (Application) Keep oil, sediment, floatables and other contaminants out of curb inlets
For Use With (Equipment) 36 to 60 in. inlets