Learn More
Teknova YPD Agar Plates with Geneticin G418-1000. 100mm plates, 20 plates. 8 pack.
SDP
Supplier:  Teknova Y11778PK
Encompass_Preferred
2% Peptone, 1% Yeast Extract, 2% Cerelose Anhydrous Dextrose (Glucose), 2% Agar,1000µg/mL Geneticin G-418 Disulfate
Catalog No. 50-238-7353
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.