Learn More
Clontech Labs 3P YPD MEDIUM 500GR
SDP
Supplier:  Clontech Labs 3P 630409
Encompass
YPD Medium 500 g blend of peptone yeast extract and dextrose in optimal proportions for growing most Saccharomyces cerevisiae strains for growing yeast cultures confirming auxotrophic phenotypes and selecting yeast transformants
Catalog No. NC9413244
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.