Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YPD (YEPD) Broth ( Powder)

Catalog No. p-4474919 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
454 g
1000 g

Rich medium for the growth of S. cerevisiae in liquid medium

  • Contains, dextrose, peptone, and yeast extract
  • pH 6.5 at 300°C
  • Bulk powder packaging
Formulation
  • 50g/L
  • Contents/liter: 20g peptone, 10g yeast extract, 20g dextrose

Yeast Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type Microbiology Media
Formulation Casein Peptone type-M 20g/L, Yeast extract 10g/L, Dextrose 20g/L
For Use With (Application) For growth of Saccharomyces cerevisiae in liquid medium
Packaging Type Bulk Powder
Quantity 1000 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.