Learn More
Clontech Labs 3P YPDA Medium | 500g
Supplier: Clontech Labs 3P 630464
YPDA yeast media pouches provide media aliquoted in highly convenient pouches. Simply empty the contents of each into your media flask/bottle, add 0.5 L of dH2O, and sterilize. No weighing of individual supplements, combining, or pH adjustment is required. All media pouch mixes are available either with agar or without (broth).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.