Learn More
Description
- Waterproof, single parameter instrument
- Automatic data storage; 16 data-sets with date and time stamp
- Large, backlit LCD display
- IP68 waterproof case; easy to hold or set on benchtop
- Innovative light shield
- Improved resolution for specific range requirements
- Auto shutoff ensures extended battery life
Chlorine Free
Range: 0.02 to 2.0mg/L
Method: DPD - Free Chlorine Reagent
Chlorine Total
Range: 0.1 to 8.0mg/L
Method: DPD - Total Chlorine Reagent
Order Info
- Dimensions: 15.5L x 7.5W x 3.8cmH (6.1 x 3 x 1.5 in.) in.
- Weight: 260g (0.5 lb.)
Certifications
CE marked IP68 (1 hour at 0.1 meter); floats
Warranty and Services
2-year
Specifications
Specifications
| Accuracy | 3% Full scale |
| Certifications/Compliance | CE |
| Data Storage | Automatic; 16 data sets with time and date stamp |
| Display | Backlit LCD |
| Dimensions (L x W x H) | 6.1 x 3 x 1.5 in. (15.5 x 7.5 x 3.8 cm) |
| IP Rating | IP68 |
| Light Source | LED |
| Type | Chlorine Colorimeter |
| Height (English) | 1.5 in. |
| Height (Metric) | 3.8 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.