Learn More
YSI™ ORP Probes for Pro Series Portable Meters
Catalog No. p-3577536 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Type:
ISE/DO/temperature probe
ORP sensor, Lab grade
Includes:
1m cable
Without cable
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Includes
15-177-461 ISE/DO/temperature probe Without cable
15-177-502 ORP sensor, Lab grade 1m cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-177-461 Supplier YSI™ Supplier No. 605102
Only null left
Add to Cart
Add to Cart
Choice of lab-grade and field models.
  • For use with Pro Series Portable Meters
  • Requires cable/probe assembly
Order Info
Lab-grade models come with cable and connector. Field model requires cable/connector assembly 15-177-466.
TRUSTED_SUSTAINABILITY
Specifications
Type ISE/DO/temperature probe
For Use With (Application) For lab-grade and field
For Use With (Equipment) Pro series meter
Includes Without cable
Probe ORP sensor