Learn More
Description
Includes
pHotoFlex pH/SET or STD/SET: pHotoFlex pH or STD colorimeter, field case with field tray, vials, pH electrode (with pH/SET), 2 each 10 mL and 2 mL syringe, empty cuvettes 16+28 mm, cleaning tissues, stand, 2 empty buffer bottles, beaker, screw driver, PC-software LSdata, and RS232 PC cable
- Large, backlit display
- Data logging enables exporting data to computer using LSdata software
- User-defined programs
- Flexible, fixed adapter with flip design
pHotoFlex STD
- For colorimeter only
- Data storage; 100 data sets
- No. of tests: 85
- For colorimeter and pH electrode
- Data storage; 1,000 data sets
- No. of tests: ∽138 tests
- Offers BNC connection
Order Info
- Dimensions : 17H x 86W x 236mm L (4.6 x 3.4 x 9.3 in.)
- Weight : 0.6kg (1.3 lb.)
Certifications
CE Marked, ETLus, cETL, IP67 waterproof
Warranty and Services
2-year
Specifications
Specifications
| Certifications/Compliance | CE, ETLus, cETL |
| Data Storage | 100 Data sets |
| Display | Backlit LCD |
| Dimensions (L x W x H) | 9.3 x 3.4 x 4.6 in. (23.6 x 8.6 x 11.7 cm) |
| IP Rating | IP67 |
| Product Line | pHotoFlex |
| Type | STD Colorimeter Set |
| Height (English) | 4.6 in. |
| Height (Metric) | 11.7 cm |
| Includes | LED filter Colorimeter, Acc. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.