Learn More
YSI™ ProODO™ Water Testing Optical DO Kit
Provides extreme durability for measuring dissolved oxygen with optical, luminescent based technology for field or lab
Manufacturer:  YSI™ 603193
 View more versions of this product
Catalog No. 09-324-502
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: ProODO meter, field cable with optical DO probe, soft-sided carrying case
Description
Description
  • Uses ODO™ technology
  • Versatile: Expanded DO range of 0-500% (0-50mg/L), backlit display and keypad, graphic display, detailed GLP data, password protection,
  • Kit includes cable with cable management kit
  • Rated to IP-67 standards even without the battery cover on
  • Built-in barometer
  • Military Spec (MS) connectors
  • Rubber overmolded case
  • Field tested, rugged cables
  • Rugged, weighted probe guard
  • 5000 data-set memory
  • Free Data Manager software
Certifications
RoHS, CE marked, WEEE, C-Tick, VCCI, IP-67, 1-meter drop test
Warranty and Services
3 Years (instrument); 2 Years (cable and probe)
Specifications
Specifications
Field
USB 2.0
24mm
32.8 ft
10m
±0.2°C
ProODO Kit
0.95 in.
7.5 in.
190mm
-20° to +70°C
ProODO meter, 10m field cable with optical DO probe, soft-sided carrying case