Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Reagent Replacement Packs for 9300 and 9500 Photometers

Catalog No. 1539912R Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Type:
Alkalinity, Total
Ammonia
Calcium Hardness
Chlorine-DPD Free
Hardness (Hardicol)
Nitrate
Nitrite-N
Phosphate LR
Sulfate
pH (Phenol Red)
Packaging:
200 Tests/Pk.
250 Tests/Pk.
Range:
0 to 0.5 mg/L
0 to 1 mg/L
0 to 20 mg/L
0 to 200 mg/L
0 to 4 mg/L
0 to 5 mg/L
0 to 500 mg/L
6.8 to 8.4 mg/L
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Range
15-399-12R Chlorine-DPD Free 250 Tests/Pk. 0 to 5 mg/L
15-399-28R Nitrate 200 Tests/Pk. 0 to 20 mg/L
15-399-2R Alkalinity, Total 250 Tests/Pk. 0 to 500 mg/L
15-399-34R Phosphate LR 200 Tests/Pk. 0 to 4 mg/L
15-399-29R Nitrite-N 250 Tests/Pk. 0 to 0.5 mg/L
15-399-33R pH (Phenol Red) 250 Tests/Pk. 6.8 to 8.4 mg/L
15-399-38R Sulfate 250 Tests/Pk. 0 to 200 mg/L
15-399-4R Ammonia 250 Tests/Pk. 0 to 1 mg/L
15-399-7R Calcium Hardness 250 Tests/Pk. 0 to 500 mg/L
15-399-18R Hardness (Hardicol) 250 Tests/Pk. 0 to 500 mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 15-399-12R Supplier YSI™ Supplier No. YAP011
Only null left
Add to Cart
Add to Cart

Specifications

Type Chlorine-DPD Free
For Use With (Equipment) 9300 and 9500 Photometer
Packaging 250 Tests/Pk.
Range 0 to 5 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.