Learn More
Medchemexpress LLC 2-Octynyladenosine | 90596-75-1 | 99.7% | 375.42 g/mol | C18H25N5O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1063035MG
Encompass_Preferred
YT 146 is a potent A2 adenosine receptor agonist used in research to stimulate intracellular cAMP signaling. It produces concentration-dependent cAMP accumulation with an EC50 of 1.5 nM and has reported cardioprotective effects. The compound is provided in small-mass research packs for laboratory applications.
  • Potent A2 adenosine receptor agonist.
  • Induces concentration-dependent cAMP accumulation (EC50 1.5 nM).
  • Reported cardioprotective effects in preclinical studies.
  • Molecular formula C18H25N5O4; molecular weight ~375.4 g/mol.
  • Supplied in small research pack sizes suitable for assay work.
Catalog No. 50-002-43769