Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YTHDF1 Mouse anti-Human, Rat, Clone: 3A2H12, Proteintech
SDP

Catalog No. p-7212816 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7154 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7154 Supplier Proteintech Group Inc Supplier No. 667451IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically recognizes and binds N6-methyladenosine (m6A)-containing RNAs. M6A is a modification present at internal sites of mRNAs and some non-coding RNAs and plays a role in the efficiency of mRNA splicing, processing and stability. Ota T., Nat. Genet. 36:40-45(2004). Deloukas P., Nature 414:865-871(2001). The MGC Project Team, Genome Res. 14:2121-2127(2004).

Specifications

Antigen YTHDF1
Applications Western Blot
Classification Monoclonal
Clone 3A2H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene YTHDF1
Gene Accession No. Q9BYJ9
Gene Alias C20orf21, DACA 1, FLJ20391, YTH domain family protein 1, YTH domain family, member 1, YTHDF1
Gene Symbols YTHDF1
Host Species Mouse
Immunogen YTHDF1 Fusion Protein Ag11633
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296467, 54915
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.