Learn More
Medchemexpress LLC Ytk-105 | 774192-20-0 | 98.8% | 257.33 g/mol | C16H19NO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW489121100MG
Encompass_Preferred
YTK-105 is a small-molecule ligand that binds the p62-ZZ domain and activates p62-dependent selective macroautophagy. Intended for research use, it is characterized by a reported purity of 98.81%, molecular formula C16H19NO2, and molecular weight 257.33 g/mol. The compound is used in autophagy-targeting studies and as a scaffold for degrader design.
  • Binds the p62-ZZ domain and modulates selective macroautophagy.
  • High reported purity suitable for biochemical and cell-based assays.
  • Characterized molecular formula and weight support reproducible experiments.
  • Suitable as a scaffold for autophagy-targeting probe and degrader design.
  • Available in milligram-scale quantities for laboratory workflows.
Catalog No. 50-002-84339
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.