Learn More
Medchemexpress LLC Ytk-105 | 774192-20-0 | 98.8% | 257.33 g·mol⁻¹ | C16H19NO2 | 50 MG

Supplier: Medchemexpress LLC HYW48912150MG
YTK-105 is a small-molecule ligand of the p62 (SQSTM1) ZZ domain that activates p62-dependent selective macroautophagy. It is intended as a research reagent for autophagy pathway studies and for use in AUTOTAC design.
- High purity: 98.8% (HPLC).
- CAS: 774192-20-0.
- Molecular formula: C16H19NO2; molecular weight: 257.33 g·mol⁻¹.
- Available in multiple pack sizes, including 50 MG.
- Useful for designing AUTOTACs and studying p62-mediated selective autophagy.
- For research use only; not for human or therapeutic use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.