Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ytk-105 | 774192-20-0 | 98.8% | 257.33 | C16H19NO2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYW4891215MG

Encompass_Preferred

YTK-105 is a small-molecule ligand of the p62 (SQSTM1) ZZ domain that activates p62-dependent selective macroautophagy. It is provided as a research tool for studying autophagy pathways and for designing AUTOTAC degraders.

  • Activates p62-dependent selective macroautophagy.
  • Useful as a probe for AUTOTAC design.
  • Molecular formula C16H19NO2; molecular weight 257.33 g·mol-1.
  • High purity (98.81%) in viscous liquid form.
  • Highly soluble in DMSO (≈175 mg/mL); ultrasonic agitation recommended.
  • Storage: pure form -20°C for long-term; in solvent -80°C short-term.

Catalog No. 50-002-84119


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.