Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ytterbium lump, sublimed dendritic, REacton™, 99.9% (REO), Thermo Scientific™

Catalog No. AA1244009
Change view
Click to view available options
Quantity:
10 g
Packaging:
Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA1244009 10 g Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1244009 Supplier Thermo Scientific Chemicals Supplier No. 1244009

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
Synonym unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
PubChem CID 23992
ChEBI CHEBI:33381
IUPAC Name ytterbium
SMILES [Yb]

Specifications

Quantity 10 g
UN Number UN1383
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Packaging Ampule
Assay (REO)
Chemical Name or Material Ytterbium lump, Sublimed dendritic
Hazard Category Danger
Hazard Statement GHS H Statement
H250-H260-H252
Precautionary Statement GHS P Statement
P210-P222-P231+P232-P280-P235+P410-P233-P422a-P420-P407-P501a
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC METALS, N.O.S.
RTECSNumber ZG1925000
TSCA TSCA
storageNote1
  • Ampouled under argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.