Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium ICP-MS Standard, 10,000 ppm Y in 5% HNO3, Ricca Chemical
SDP

Catalog No. MSY10KN100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSY10KN-100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSY10KN-100 Supplier Ricca Chemical Company Supplier No. MSY10KN100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1314-36-9 , 7697-37-2 , 7732-18-5
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm Y
CAS Min % 85.0, 11.83, 1.08
CAS Max % 89.0, 12.31, 1.13
Equivalence 1 mL = 10 mg Y
Physical Form Liquid
Color Colorless
Quantity 100 mL
Chemical Name or Material Yttrium ICP-MS Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.