Yttrium(III) acetate tetrahydrate, 99.99% (REO), Alfa Aesar™, Reacton™

Manufacturer: Alfa Aesar 1118622

Quantity 100g

 View more versions of this product

Catalog No. AA1118622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85949-60-6
    Physical Form Crystalline Aggregates
    Assay Percent Range 99.99% (REO)
    Molecular Formula C6H9O6Y·4H2O
    Formula Weight 338.10; 266.04 (anhydrous)
    MDL Number MFCD00013048
    Chemical Name or Material Yttrium(III) acetate tetrahydrate

    Specifications

    Formula Weight 338.10; 266.04 (anhydrous)
    Quantity 100g
    EINECS Number 245-612-0
    Physical Form Crystalline Aggregates
    Density 1.5g/mL
    Assay Percent Range 99.99% (REO)
    Molecular Formula C6H9O6Y·4H2O
    MDL Number MFCD00013048
    CAS 85949-60-6
    Chemical Name or Material Yttrium(III) acetate tetrahydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit