Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) carbonate hydrate, 99.9% (REO)

Catalog No. AA4065118
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4065118 50 g
AA4065130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4065118 Supplier Thermo Scientific Chemicals Supplier No. 04065118
Add to Cart

CAS: 38245-39-5 | C3O9Y2 | 357.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38245-39-5
Molecular Formula C3O9Y2
Molecular Weight (g/mol) 357.84
MDL Number MFCD00149943
InChI Key QVOIJBIQBYRBCF-UHFFFAOYSA-H
IUPAC Name diyttrium(3+) tricarbonate
SMILES [Y+3].[Y+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Physical Form Powder
Quantity 50 g
Merck Index 14,10107
Formula Weight 357.84 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Yttrium(III) carbonate hydrate
EINECSNumber 209-119-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.